Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibasic Sodium Phosphate, Dried, USP, 98-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7558-79-4
HNa2O4P
141.96
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate
[Na+].[Na+].OP([O-])([O-])=O
7558-79-4
HNa2O4P
141.96
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate
[Na+].[Na+].OP([O-])([O-])=O

SO138, 7558-79-4, Na2HPO4

Spectrum™ Dibasic Sodium Phosphate, Dried, USP is used as a saline laxative to treat constipation or to clean the bowel before a colonoscopy. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 16 ppm
Percent Purity 98 to 100.5%
Chloride 0.0006
Insoluble Matter 0.004
Loss on Drying 0.05
Packaging Poly Bottle
Sulfate 0.002
Quantity 4 x 2.5 kg