Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibasic Sodium Phosphate, Heptahydrate, Crystal, USP, 98-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7782-85-6
H17Na2O11P
270.08
PYLIXCKOHOHGKQ-UHFFFAOYSA-N
disodium phosphoric acid heptahydrate
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O
7782-85-6
H17Na2O11P
270.08
PYLIXCKOHOHGKQ-UHFFFAOYSA-N
disodium phosphoric acid heptahydrate
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O

SO140, 7782-85-6, Na2HPO4•7H2O

Spectrum™ Dibasic Sodium Phosphate, Heptahydrate, Crystal, USP is used as a saline laxative to treat constipation or to clean the bowel before a colonoscopy. Spectrum Chemical USP products, including all USP Sodium Phosphate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 16 ppm
Percent Purity 98 to 100.5%
Chloride 0.0006
Insoluble Matter 0.004
Loss on Drying 43.0-50%
Packaging Poly Bottle
Sulfate 0.002
Quantity 2.5 kg