Alfa Aesar™ Dibenzenechromium, C 69.2%, H 5.8%

Manufacturer: Alfa Aesar™ 2413902

Quantity 0.5g

 View more versions of this product

Catalog No. AA2413902

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1271-54-1
    Linear Formula (C6H6)2Cr
    Formula Weight 208.22
    MDL Number MFCD00015485
    UN Number UN3391
    Synonym Chromium dibenzene
    Chemical Name or Material Dibenzenechromium

    Specifications

    Formula Weight 208.22
    Health Hazard 3 GHS P Statement
    P210-P222-P280-P370+P378a-P422a-P420
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store contents under inert gas.
    Store away from other materials.
    CAS 1271-54-1
    Health Hazard 1 Danger
    Physical Form Crystalline
    Linear Formula (C6H6)2Cr
    UN Number UN3391
    Color Black
    Sensitivity Air & Moisture Sensitive
    MDL Number MFCD00015485
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC
    Melting Point 260° to 265°C
    Quantity 0.5g
    Concentration C 69.2%, H 5.8%
    Synonym Chromium dibenzene
    Health Hazard 2 GHS H Statement
    H250-H252
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    RTECS Number GB5850000
    Chemical Name or Material Dibenzenechromium

    GHS H Statement
    H250-H252
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.

    GHS P Statement
    P210-P222-P280-P370+P378a-P422a-P420
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store contents under inert gas.
    Store away from other materials.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit