Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzenesulfonimide 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
2618-96-4
C12H11NO4S2
297.343
MFCD00025019
OVQABVAKPIYHIG-UHFFFAOYSA-N
dibenzenesulfonimide,benzenesulfonamide, n-phenylsulfonyl,dibenzenesulfonamide,bis phenylsulfonyl amine,n-phenylsulfonyl benzenesulfonamide,dibenzolsulfimid,dibenzolsulfimide,diphenylsulfonimide,benzenesulfonic acid imide,n-benzenesulfonyl benzenesulfonamide
75671
N-(benzenesulfonyl)benzenesulfonamide
C1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=CC=C2
2618-96-4
C12H11NO4S2
297.343
MFCD00025019
OVQABVAKPIYHIG-UHFFFAOYSA-N
dibenzenesulfonimide,benzenesulfonamide, n-phenylsulfonyl,dibenzenesulfonamide,bis phenylsulfonyl amine,n-phenylsulfonyl benzenesulfonamide,dibenzolsulfimid,dibenzolsulfimide,diphenylsulfonimide,benzenesulfonic acid imide,n-benzenesulfonyl benzenesulfonamide
75671
N-(benzenesulfonyl)benzenesulfonamide
C1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=CC=C2
Color White-Yellow
Melting Point 158°C
Quantity 25g
Formula Weight 297.34
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Dibenzenesulfonimide

EINECSNumber : (3)-3133

TSCA : Yes