Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzo-30-crown-10, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
17455-25-3
C28H40O10
536.618
MFCD00054624
MXCSCGGRLMRZMF-UHFFFAOYSA-N
dibenzo-30-crown-10,dibenzo-30-crown 10-ether,dibenzo-30-crown,dibenzo-30-crown-10-ether,2,3,17,18-dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriaconta-2,17-diene
87120
C1COCCOC2=CC=CC=C2OCCOCCOCCOCCOC3=CC=CC=C3OCCOCCO1
17455-25-3
C28H40O10
536.618
MFCD00054624
MXCSCGGRLMRZMF-UHFFFAOYSA-N
dibenzo-30-crown-10,dibenzo-30-crown 10-ether,dibenzo-30-crown,dibenzo-30-crown-10-ether,2,3,17,18-dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriaconta-2,17-diene
87120
C1COCCOC2=CC=CC=C2OCCOCCOCCOCCOC3=CC=CC=C3OCCOCCO1
Chelation/Complexation reagent
Packaging Glass bottle
Color White
Quantity 250mg
Melting Point 104°C to 108°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 536.62
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Dibenzo-30-crown-10, 98%

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.