Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dibenzofuran-2-carboxaldehyde, 98%

Click to view available options
Quantity:
250mg
1g
5g
5397-82-0
C13H8O2
196.205
MFCD00136272
OVJMIWIVPWPZMN-UHFFFAOYSA-N
dibenzo b,d furan-2-carbaldehyde,2-dibenzofurancarboxaldehyde,dibenzofuran-2-carboxaldehyde,dibenzo b,d furan-2-carboxaldehyde,8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde,2-dibenzofurancarbaldehyde,dibenzo b furan-2-carbaldehyde,benzo b benzofuran-2-carboxaldehyde,8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene-4-carbaldehyde,8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde
220843
dibenzofuran-2-carbaldehyde
C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)C=O
5397-82-0
C13H8O2
196.205
MFCD00136272
OVJMIWIVPWPZMN-UHFFFAOYSA-N
dibenzo b,d furan-2-carbaldehyde,2-dibenzofurancarboxaldehyde,dibenzofuran-2-carboxaldehyde,dibenzo b,d furan-2-carboxaldehyde,8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde,2-dibenzofurancarbaldehyde,dibenzo b furan-2-carbaldehyde,benzo b benzofuran-2-carboxaldehyde,8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene-4-carbaldehyde,8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde
220843
dibenzofuran-2-carbaldehyde
C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)C=O
Applications
Used as an pharmaceutical intermediate.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 250mg
Color White
Melting Point 74°C
Sensitivity Air Sensitive
Beilstein 141044
Formula Weight 196.2
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Dibenzofuran-2-carboxaldehyde

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger