Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzosuberenol, 96%, Thermo Scientific Chemicals

Catalog Number p-7043684
Click to view available options
Quantity:
5 g
10354-00-4
C15H12O
208.26
MFCD00003586
SRIISEYIFDTFRZ-UHFFFAOYSA-N
dibenzosuberenol, 5h-dibenzo a,d 7 annulen-5-ol, 5h-dibenzo a,d cyclohepten-5-ol, unii-s9t01jbh4a, s9t01jbh4a, 5h-dibenzo a,d-1-cyclohepten-5-ol, dibenzo b,f cyclohepten-1-ol, 5h-dibenzo-a,d-cyclohepten-5-ol, 5-hydrodibenzo a,d 7 annulen-5-ol
82577
11H-dibenzo[1,2-a
C1=CC=C2C(C3=CC=CC=C3C=CC2=C1)O
10354-00-4
C15H12O
208.26
MFCD00003586
SRIISEYIFDTFRZ-UHFFFAOYSA-N
dibenzosuberenol, 5h-dibenzo a,d 7 annulen-5-ol, 5h-dibenzo a,d cyclohepten-5-ol, unii-s9t01jbh4a, s9t01jbh4a, 5h-dibenzo a,d-1-cyclohepten-5-ol, dibenzo b,f cyclohepten-1-ol, 5h-dibenzo-a,d-cyclohepten-5-ol, 5-hydrodibenzo a,d 7 annulen-5-ol
82577
11H-dibenzo[1,2-a
C1=CC=C2C(C3=CC=CC=C3C=CC2=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 125°C to 127°C
Quantity 5 g
Beilstein 1957458
Formula Weight 208.26
Percent Purity 96%
Chemical Name or Material Dibenzosuberenol
EINECSNumber 233-774-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit