Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dibenzosuberenone, 97%

Click to view available options
Quantity:
5g
25g
2222-33-5
C15H10O
206.244
MFCD00003588
SNVTZAIYUGUKNI-UHFFFAOYSA-N
dibenzosuberenone,5-dibenzosuberenone,5h-dibenzo a,d cyclohepten-5-one,5h-dibenzo a,d 7 annulen-5-one,dibenzo a,d cyclohepten-5-one,2,3:6,7-dibenzotropone,2,3:6,7-dibenzosuberen-4-on-1,5h-dibenzo a,d cycloheptenone
16679
dibenzo[1,2-a
C1=CC=C2C(=C1)C=CC3=CC=CC=C3C2=O
2222-33-5
C15H10O
206.244
MFCD00003588
SNVTZAIYUGUKNI-UHFFFAOYSA-N
dibenzosuberenone,5-dibenzosuberenone,5h-dibenzo a,d cyclohepten-5-one,5h-dibenzo a,d 7 annulen-5-one,dibenzo a,d cyclohepten-5-one,2,3:6,7-dibenzotropone,2,3:6,7-dibenzosuberen-4-on-1,5h-dibenzo a,d cycloheptenone
16679
dibenzo[1,2-a
C1=CC=C2C(=C1)C=CC3=CC=CC=C3C2=O
Applications
Dibenzosuberenone can be used in the synthesis of a series of potent antidepressants.

Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.
Quantity 5g
Melting Point 88°C to 89°C
Beilstein 743171
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material Dibenzosuberenone

RTECSNumber : HP1147500

TSCA : TSCA