Alfa Aesar™ Dibenzosuberenone, 97%

Manufacturer: Alfa Aesar™ B2372722

Quantity 100g

 View more versions of this product

Catalog No. AAB2372722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2222-33-5
    Molecular Formula C15H10O
    Formula Weight 206.24
    MDL Number MFCD00003588
    Beilstein 743171
    Synonym 5H-Dibenzo[a,d]cyclohepten-5-one
    Chemical Name or Material Dibenzosuberenone

    Specifications

    Formula Weight 206.24
    TSCA TSCA
    Melting Point 88° to 89°C
    Quantity 100g
    Molecular Formula C15H10O
    MDL Number MFCD00003588
    CAS 2222-33-5
    Beilstein 743171
    Synonym 5H-Dibenzo[a,d]cyclohepten-5-one
    RTECS Number HP1147500
    Chemical Name or Material Dibenzosuberenone
    Assay 97%

    Description

    Applications
    Dibenzosuberenone can be used in the synthesis of a series of potent antidepressants.

    Solubility
    Soluble in chloroform, ethyl acetate. Insoluble in water.

    Notes
    Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit