Alfa Aesar™ Dibenzosuberol, 98%

Manufacturer: Alfa Aesar™ B2235422

Quantity 100g

 View more versions of this product

Catalog No. AAB2235422

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1210-34-0
    Molecular Formula C15H14O
    Formula Weight 210.28
    MDL Number MFCD00003593
    Beilstein 520430
    Synonym 10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ol
    Chemical Name or Material Dibenzosuberol

    Specifications

    Formula Weight 210.28
    Melting Point 90° to 94°C
    Boiling Point 166° to 169°C (1mmHg)
    Molecular Formula C15H14O
    MDL Number MFCD00003593
    CAS 1210-34-0
    Beilstein 520430
    Quantity 100g
    Synonym 10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ol
    RTECS Number HP1100000
    Chemical Name or Material Dibenzosuberol
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit