Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dibenzothiophene-2-boronic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
668983-97-9
C12H9BO2S
228.072
MFCD01318982
CSLSCVHILGCSTE-UHFFFAOYSA-N
dibenzo b,d thiophen-2-ylboronic acid,dibenzothiophene-2-boronic acid,dibenzo b,d thien-2-ylboronic acid,dibenzothiophen-2-yl-dihydroxy-borane,boronic acid, 2-dibenzothienyl,boronic acid, b-2-dibenzothienyl,dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride,dibenzo b,d thien-2-ylboronic acid saltdata: free,8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid,ksc352o5l
2794660
dibenzothiophen-2-ylboronic acid
B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O
668983-97-9
C12H9BO2S
228.072
MFCD01318982
CSLSCVHILGCSTE-UHFFFAOYSA-N
dibenzo b,d thiophen-2-ylboronic acid,dibenzothiophene-2-boronic acid,dibenzo b,d thien-2-ylboronic acid,dibenzothiophen-2-yl-dihydroxy-borane,boronic acid, 2-dibenzothienyl,boronic acid, b-2-dibenzothienyl,dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride,dibenzo b,d thien-2-ylboronic acid saltdata: free,8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid,ksc352o5l
2794660
dibenzothiophen-2-ylboronic acid
B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O
Quantity 1g
Color White
Beilstein 15124
Formula Weight 228.07
Physical Form Powder
Percent Purity 97%
Chemical Name or Material Dibenzothiophene-2-boronic acid

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C