Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzothiophene-4-boronic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
108847-20-7
C12H9BO2S
228.08
GOXNHPQCCUVWRO-UHFFFAOYSA-N
dibenzothiophene-4-boronic acid, 4-dibenzothiopheneboronic acid, dibenzo b,d thiophen-4-ylboronic acid, 4-dibenzothienylboronic acid, dibenzo b,d thiophene-4-boronic acid, boronic acid, 4-dibenzothienyl, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid, pubchem7386, 4-dibenzothiopheneboronicacid
2734329
dibenzothiophen-4-ylboronic acid
B(C1=C2C(=CC=C1)C3=CC=CC=C3S2)(O)O
108847-20-7
C12H9BO2S
228.08
GOXNHPQCCUVWRO-UHFFFAOYSA-N
dibenzothiophene-4-boronic acid, 4-dibenzothiopheneboronic acid, dibenzo b,d thiophen-4-ylboronic acid, 4-dibenzothienylboronic acid, dibenzo b,d thiophene-4-boronic acid, boronic acid, 4-dibenzothienyl, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid, pubchem7386, 4-dibenzothiopheneboronicacid
2734329
dibenzothiophen-4-ylboronic acid
B(C1=C2C(=CC=C1)C3=CC=CC=C3S2)(O)O
Infrared Spectrum Authentic
Melting Point 285.0°C to 290.0°C
Quantity 5g
Color White
Assay Percent Range 94% min. (HPLC)
Formula Weight 228.08
Physical Form Powder
Percent Purity 95%
Chemical Name or Material Dibenzothiophene-4-boronic acid

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER

GHS Signal Word: Warning