Alfa Aesar™ Dibenzothiophene, 98%

Manufacturer: Alfa Aesar™ A1228814

Quantity 25g

 View more versions of this product

Catalog No. AAA1228814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 132-65-0
    Molecular Formula C12H8S
    Formula Weight 184.25
    MDL Number MFCD00004969
    UN Number UN3077
    Beilstein 121101
    Synonym Diphenylene sulfide
    Chemical Name or Material Dibenzothiophene

    Specifications

    Formula Weight 184.25
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Melting Point 97° to 100°C
    Quantity 25g
    MDL Number MFCD00004969
    CAS 132-65-0
    UN Number UN3077
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Boiling Point 332° to 333°C
    Molecular Formula C12H8S
    Beilstein 121101
    Synonym Diphenylene sulfide
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Flash Point 170°C (338°F)
    RTECS Number HQ3490550
    Chemical Name or Material Dibenzothiophene
    Assay 98%

    Description

    Applications
    Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

    Solubility
    Soluble in water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit