Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+)-Dibenzoyl-D-tartaric acid, anhydrous, 99%

Click to view available options
Quantity:
10g
50g
250g
17026-42-5
C18H14O8
358.302
MFCD00063222
YONLFQNRGZXBBF-KBPBESRZSA-N
dibenzoyl-d-tartaric acid,+-dibenzoyl-d-tartaric acid,2s,3s-2,3-bis benzoyloxy succinic acid,d-dibenzoyl tartaric acid,butanedioic acid, 2,3-bis benzoyloxy-, 2s,3s,+-o,o-dibenzoyltartaric acid,2s,3s-+-dibenzoyltartaric acid,+-dibenzoyltartaric acid,+-dibenzoyl tartaric acid
1550213
(2S,3S)-2,3-dibenzoyloxybutanedioic acid
C1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=CC=C2)C(=O)O
17026-42-5
C18H14O8
358.302
MFCD00063222
YONLFQNRGZXBBF-KBPBESRZSA-N
dibenzoyl-d-tartaric acid,+-dibenzoyl-d-tartaric acid,2s,3s-2,3-bis benzoyloxy succinic acid,d-dibenzoyl tartaric acid,butanedioic acid, 2,3-bis benzoyloxy-, 2s,3s,+-o,o-dibenzoyltartaric acid,2s,3s-+-dibenzoyltartaric acid,+-dibenzoyltartaric acid,+-dibenzoyl tartaric acid
1550213
(2S,3S)-2,3-dibenzoyloxybutanedioic acid
C1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=CC=C2)C(=O)O
Applications
For chiral resolution(+)-Dibenzoyl-D-tartaric acid is used as a reagent in the preparation of chiral salts. It also serves as a reagent for chiral resolution of amino compounds.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Beilstein 2227343
Formula Weight 358.31
Percent Purity 99%
Melting Point 154°C to 156°C
Quantity 50g
Optical Rotation +120° (c=5 in Methanol)
Chemical Name or Material (+)-Dibenzoyl-D-tartaric acid, anhydrous

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA