Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (-)-Dibenzoyl-L-tartaric acid, anhydrous, 99%

Click to view available options
Quantity:
25g
100g
500g
2743-38-6
C18H14O8
358.302
MFCD00003074
YONLFQNRGZXBBF-ZIAGYGMSSA-N
dibenzoyl-l-tartaric acid,2r,3r-2,3-bis benzoyloxy succinic acid,dibenzoyltartaric acid,--dibenzoyl-l-tartaric acid,dibenzoyl-l-tartaricacid,l-dibenzoyl tartaric acid,butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r,unii-3q14b666vm,2,3-bis benzoyloxy tartaric acid,o,o-dibenzoyl-+-tartaric acid
75969
(2R,3R)-2,3-dibenzoyloxybutanedioic acid
C1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=CC=C2)C(=O)O
2743-38-6
C18H14O8
358.302
MFCD00003074
YONLFQNRGZXBBF-ZIAGYGMSSA-N
dibenzoyl-l-tartaric acid,2r,3r-2,3-bis benzoyloxy succinic acid,dibenzoyltartaric acid,--dibenzoyl-l-tartaric acid,dibenzoyl-l-tartaricacid,l-dibenzoyl tartaric acid,butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r,unii-3q14b666vm,2,3-bis benzoyloxy tartaric acid,o,o-dibenzoyl-+-tartaric acid
75969
(2R,3R)-2,3-dibenzoyloxybutanedioic acid
C1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=CC=C2)C(=O)O
Applications
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
It is sensitive to moisture. Incompatible with oxidizing agents.
Solubility Information Slightly soluble in water.
Formula Weight 358.31
Percent Purity 99%
Melting Point 154°C to 156°C
Quantity 500g
Sensitivity Hygroscopic
Chemical Name or Material (-)-Dibenzoyl-L-tartaric acid, anhydrous

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 220-374-0

RTECSNumber : WW8070000

TSCA : Yes

Recommended Storage : Ambient temperatures