Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzoyl peroxide, 75%, remainder water, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
Packaging:
Plastic bottle
94-36-0,7732-18-5
C14H10O4
242.23
MFCD00003071
OMPJBNCRMGITSC-UHFFFAOYSA-N
benzoyl peroxide,dibenzoyl peroxide,peroxide, dibenzoyl,benzoperoxide,benzoyl superoxide,acetoxyl,lucidol,benoxyl,panoxyl,benzol peroxide
7187
CHEBI:82405
benzoyl benzenecarboperoxoate
C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2
94-36-0,7732-18-5
C14H10O4
242.23
MFCD00003071
OMPJBNCRMGITSC-UHFFFAOYSA-N
benzoyl peroxide,dibenzoyl peroxide,peroxide, dibenzoyl,benzoperoxide,benzoyl superoxide,acetoxyl,lucidol,benoxyl,panoxyl,benzol peroxide
7187
CHEBI:82405
benzoyl benzenecarboperoxoate
C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2
Radical initiators
Color White
Quantity 5g
Packaging Plastic bottle
Melting Point 104.0°C to 106.0°C
Linear Formula (C6H5CO)2O2
Beilstein 09, 179
Fieser 01,196; 04,122; 05,182; 06,160; 07,89; 12,157; 15,146; 17,367
Merck Index 15, 1119
Solubility Information Solubility in water: insoluble. Other solubilities: sparingly soluble in alcohol,soluble in benzene,chloroform,ether and acetone,1g/40 mL carbon disulfide,1g/40 mL olive oil
Formula Weight 242.23
Physical Form Wet Granular Powder
Percent Purity 75%
Chemical Name or Material Dibenzoyl peroxide, 75%, remainder water
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Heating may cause a fire.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash

GHS Signal Word: Danger

EINECSNumber : 202-327-6

RTECSNumber : DM8575000

TSCA : TSCA