Alfa Aesar™ Dibenzyl carbonate, 98%

Manufacturer: Alfa Aesar™ A1146430

Quantity 250g

 View more versions of this product

Catalog No. AAA1146430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3459-92-5
    Molecular Formula C15H14O3
    Formula Weight 242.27
    MDL Number MFCD00014436
    Beilstein 1882864
    Synonym Benzyl carbonate; Carbonic acid dibenzyl ester
    Chemical Name or Material Dibenzyl carbonate

    Specifications

    Formula Weight 242.27
    Boiling Point 150° to 155°C (0.8mmHg)
    Quantity 250g
    Flash Point >110°C (230°F)
    MDL Number MFCD00014436
    CAS 3459-92-5
    Synonym Benzyl carbonate; Carbonic acid dibenzyl ester
    TSCA TSCA
    Molecular Formula C15H14O3
    Beilstein 1882864
    Melting Point 27° to 30°C
    Chemical Name or Material Dibenzyl carbonate
    Assay 98%

    Description

    Applications
    Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.

    Solubility
    Sparingly Soluble (0.099 g/L) (25°C).

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit