Alfa Aesar™ Dibenzyl diethylphosphoramidite, 90+%

Manufacturer: Alfa Aesar™ B2561903

Quantity 1g

 View more versions of this product

Catalog No. AAB2561903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67746-43-4
    Molecular Formula C18H24NO2P
    Formula Weight 317.37
    MDL Number MFCD00191987
    Synonym DDP
    Chemical Name or Material Dibenzyl diethylphosphoramidite

    Specifications

    Formula Weight 317.37
    Quantity 1g
    Flash Point 73°C (163°F)
    Molecular Formula C18H24NO2P
    MDL Number MFCD00191987
    Synonym DDP
    Health Hazard 1 Warning
    CAS 67746-43-4
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Chemical Name or Material Dibenzyl diethylphosphoramidite
    Assay ≥90%

    Description

    Applications
    Dibenzyl N,N-diethylphosphoramidite may be used as phosphitylating reagent in the synthesis of glycosyl phosphites and phosphates, in the preparation of 6-dibenzylphosphate-1,3,5-tri-O-PMB-myo-inositol, for phosphorylating the alcohols.

    Solubility
    Soluble in chloroform.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit