Learn More
Dibenzyl malonate, 95%, Thermo Scientific Chemicals
Description
Dibenzyl malonate is used in the preparation of tetraethyl 3,3-bis(benzyloxycarbonyl)propylene bisphosphonate. It was also used in the preparation of benzyl umbelliferone-3-carboxylate via Knoevenagel condensation with 2,4-dihydroxybezaldehyde. It is a useful alternative to Diethyl malonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dibenzyl malonate is used in the preparation of tetraethyl 3,3-bis(benzyloxycarbonyl)propylene bisphosphonate. It was also used in the preparation of benzyl umbelliferone-3-carboxylate via Knoevenagel condensation with 2,4-dihydroxybezaldehyde. It is a useful alternative to Diethyl malonate.
Solubility
Sparingly soluble in water; miscible in all proportions with ether and alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Specifications
Density | 1.137 |
Boiling Point | 188°C (0.2 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.544 |
Quantity | 10 g |
Beilstein | 1998264 |
Solubility Information | Sparingly soluble in water; miscible in all proportions with ether and alcohol. |
Formula Weight | 284.31 |
Percent Purity | 95% |
Chemical Name or Material | Dibenzyl malonate |
Safety and Handling
EINECSNumber | 239-099-2 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.