Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzyl oxalate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
7579-36-4
C16H14O4
270.284
MFCD00426620
ZYZXGWGQYNTGAU-UHFFFAOYSA-N
dibenzyl oxylate,oxalic acid, dibenzyl ester,benzyl oxalate,ethanedioic acid, bis phenylmethyl ester,unii-9a31q81ddc,ethanedioic acid, 1,2-bis phenylmethyl ester,dibenzyloxalat,dibenzyloxalate,dibenzyl-oxalate,dibenzyl ethanedioate
82058
dibenzyl oxalate
C1=CC=C(C=C1)COC(=O)C(=O)OCC2=CC=CC=C2
7579-36-4
C16H14O4
270.284
MFCD00426620
ZYZXGWGQYNTGAU-UHFFFAOYSA-N
dibenzyl oxylate,oxalic acid, dibenzyl ester,benzyl oxalate,ethanedioic acid, bis phenylmethyl ester,unii-9a31q81ddc,ethanedioic acid, 1,2-bis phenylmethyl ester,dibenzyloxalat,dibenzyloxalate,dibenzyl-oxalate,dibenzyl ethanedioate
82058
dibenzyl oxalate
C1=CC=C(C=C1)COC(=O)C(=O)OCC2=CC=CC=C2
Melting Point 75°C to 78°C
Packaging Glass Bottle
Color White
Quantity 25g
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula C6H5CH2OCOCOOCH2C6H5
Formula Weight 270.28
Physical Form Flaked Crystalline Solid
Percent Purity ≥96.0%
Chemical Name or Material Dibenzyl oxalate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.