Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dibenzyl sulfide, 99%

Manufacturer:  Alfa Aesar™ B2233714

Quantity 25g

 View more versions of this product

Catalog No. AAB2233714


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

538-74-9
214.33
1911157
Benzyl sulfide
(C6H5CH2)2S
MFCD00004784
14,1145
Dibenzyl sulfide

Specifications

538-74-9
25g
(C6H5CH2)2S
MFCD00004784
14,1145
Dibenzyl sulfide
47° to 50°C
>110°C (230°F)
214.33
1911157
Benzyl sulfide
99%

Safety and Handling

SDS & Safety Documents

RTECSNumber : DC0512500

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit