Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dibenzyl sulfide, 99%

Click to view available options
Quantity:
25g
100g
538-74-9
C14H14S
214.326
MFCD00004784
LUFPJJNWMYZRQE-UHFFFAOYSA-N
dibenzyl sulfide,benzyl sulfide,dibenzyl sulphide,benzyl thioether,benzyl monosulfide,dibenzyl monosulfide,dibenzyl thioether,sulfide, dibenzyl,1,3-diphenyl-2-thiapropane,benzene, 1,1'-thiobis methylene bis
10867
benzylsulfanylmethylbenzene
C1=CC=C(C=C1)CSCC2=CC=CC=C2
538-74-9
C14H14S
214.326
MFCD00004784
LUFPJJNWMYZRQE-UHFFFAOYSA-N
dibenzyl sulfide,benzyl sulfide,dibenzyl sulphide,benzyl thioether,benzyl monosulfide,dibenzyl monosulfide,dibenzyl thioether,sulfide, dibenzyl,1,3-diphenyl-2-thiapropane,benzene, 1,1'-thiobis methylene bis
10867
benzylsulfanylmethylbenzene
C1=CC=C(C=C1)CSCC2=CC=CC=C2
Quantity 100g
Flash Point >110°C (230°F)
Melting Point 47°C to 50°C
Beilstein 1911157
Merck Index 14,1145
Formula Weight 214.33
Percent Purity 99%
Chemical Name or Material Dibenzyl sulfide

RTECSNumber : DC0512500

TSCA : TSCA