Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dibenzylamine, 98%

Manufacturer:  Alfa Aesar™ A115540I

Quantity 5000g

 View more versions of this product

Catalog No. AAA115540I


  Check Availability
Add to cart

Specifications

Description

Applications
Dibenzylamine is used to prepare rubber accelerator for the vulcanization process and reaction-stoppers. Dibenzylamine type accelerators reduce nitrosamine production in the compounding process. It is also used to produce dibenzyl-nitroso-amine.

Solubility
Soluble in water(0.4g/L).

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

103-49-1
197.28
UN2735
14,3011
C14H15N
MFCD00004770
909664
Dibenzylamine

Specifications

103-49-1
300°C
143°C (289°F)
Weak
MFCD00004770
UN2735
14,3011
98%
−26°C
5000g
C14H15N
197.28
8.9
909664
Dibenzylamine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit