Alfa Aesar™ Dibromo[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II)

Manufacturer: Alfa Aesar™ 4669603

Quantity 1g

 View more versions of this product

Catalog No. AA4669603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1154043-20-5
    Formula Weight 684.55
    MDL Number MFCD14155728
    Synonym PdBr2(dippf); [1,1'-Bis(diisopropylphosphino)ferrocene]dibromopalladium(II)
    Chemical Name or Material Dibromo[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II)

    Specifications

    Formula Weight 684.55
    Color Red
    Physical Form Crystalline Solid
    MDL Number MFCD14155728
    CAS 1154043-20-5
    Quantity 1g
    Synonym PdBr2(dippf); [1,1'-Bis(diisopropylphosphino)ferrocene]dibromopalladium(II)
    Chemical Name or Material Dibromo[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit