Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dibromo(1,5-cyclooctadiene)platinum(II), Pt 41.6% min

Click to view available options
Quantity:
1g
5g
12145-48-1
C8H12Br2Pt
463.076
MFCD00058724
HFLCCHAOKSABAA-PHFPKPIQSA-L
dibromo 1,5-cyclooctadiene platinum ii,c8h12.br2pt,1z,5z-cycloocta-1,5-diene; dibromoplatinum,1,5-cyclooctadiene platinum ii bromide,dibromo cycloocta-1,5-diene platinum ii,1,5-cyclooctadiene, z,z-; dibromoplatinum
92135605
(1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dibromide
C1CC=CCCC=C1.[Br-].[Br-].[Pt+2]
12145-48-1
C8H12Br2Pt
463.076
MFCD00058724
HFLCCHAOKSABAA-PHFPKPIQSA-L
dibromo 1,5-cyclooctadiene platinum ii,c8h12.br2pt,1z,5z-cycloocta-1,5-diene; dibromoplatinum,1,5-cyclooctadiene platinum ii bromide,dibromo cycloocta-1,5-diene platinum ii,1,5-cyclooctadiene, z,z-; dibromoplatinum
92135605
(1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dibromide
C1CC=CCCC=C1.[Br-].[Br-].[Pt+2]
Concentration or Composition (by Analyte or Components) Pt 41.6% min.
Quantity 5g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 463.09
Physical Form Powder
Chemical Name or Material Dibromo(1,5-cyclooctadiene)platinum(II)

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures