Learn More
Dibromo-tyrosine Mouse anti-Chemical, Clone: 6G3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527565
Catalog No. PIMA527565
Add to cart
Description
Description
Dibromotyrosine is an antithyroid preparation and a derivative of the natural amino acid tyrosine.
Specifications
Specifications
Dibromo-tyrosine
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
3,5-Dibromo-tyrosine, 3,5-dibromotyrosine, Dibromotyrosine, Dibromo-tyrosine, DiBrY, Dibromotyrosine (DiBrY), Dibromo-Tyrosine (DiBrY), Dibromo-tyrosine [DiBrY]
Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH)
Protein G
RUO
Antibody
Monoclonal
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
6G3
Unconjugated
Liquid
Mouse
IgG1
100 μg
-20°C
Primary
Chemical