Learn More
Dibromo-tyrosine Mouse anti-Non-species specific, Biotin, Clone: 9F12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259351B
Catalog No. NBP259351B
Add to cart
Description
Description
Dibromo-tyrosine Monoclonal antibody specifically detects Dibromo-tyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Dibromo-tyrosine
9F12
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
Mouse
IgG1
100 ul
Primary
Specific for 3,5-Dibromo-tyrosine. Does not cross-react with Nitrotyrosine.