Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibromo-tyrosine Mouse anti-Non-species specific, Clone: 6G3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259350

Catalog No. NBP259350


Add to cart

Description

Description

Dibromo-tyrosine Monoclonal antibody specifically detects Dibromo-tyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Dot Blot.
Specifications

Specifications

Dibromo-tyrosine
6G3
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Dot Blot
Purified
Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
Protein G purified
RUO
Primary
Specific for 3,5-Dibromo-tyrosine. Does not cross-react with Nitrotyrosine.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot
Unconjugated
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Mouse
IgG1
100 ug
Store at -20C.
Monoclonal
Non-species specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only