Learn More
Dibromo-tyrosine Mouse anti-Non-species specific, DyLight 405, Clone: 9F12, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259351V
Description
Dibromo-tyrosine Monoclonal specifically detects Dibromo-tyrosine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Dibromo-tyrosine | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
100 μL | |
Specific for 3,5-Dibromo-tyrosine. Does not cross-react with Nitrotyrosine. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
9F12 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein G purified | |
Primary | |
Nonspecies specific | |