Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibromo-tyrosine Mouse, Streptavidin, Clone:6G3, Abnova™

Mouse monoclonal antibody raised against synthetic Dibromo-tyrosine (DiBrY).

Manufacturer:  Abnova Corporation MAB17390

Catalog No. 89-334-258


Add to cart

Specifications

Specifications

Dibromo-tyrosine
6G3
ELISA (1:1000)
Immunocytochemistry (1:50)
Immunofluorescence (1:50)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Liquid
Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH).
100μg
Antibody
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Streptavidin
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Mouse
IgG1
Store at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit