Alfa Aesar™ Dibromobis(triphenylphosphine)nickel(II), 99%

As a polymerization catalyst

Manufacturer: Alfa Aesar™ 4219214

Quantity 25g

 View more versions of this product

Catalog No. AA4219214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14126-37-5
    Formula Weight 743.12
    MDL Number MFCD00015865
    Synonym Bis(triphenylphosphine)nickel(II) bromide; Bis(triphenylphosphine)dibromonickel(II)
    Chemical Name or Material Dibromobis(triphenylphosphine)nickel(II)

    Specifications

    Formula Weight 743.12
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 219° to 223°C
    Physical Form Powder
    Quantity 25g
    Odor Odorless
    MDL Number MFCD00015865
    Synonym Bis(triphenylphosphine)nickel(II) bromide; Bis(triphenylphosphine)dibromonickel(II)
    Health Hazard 1 Danger
    CAS 14126-37-5
    Health Hazard 2 GHS H Statement
    H350-H317
    May cause cancer.
    May cause an allergic skin reaction.
    Chemical Name or Material Dibromobis(triphenylphosphine)nickel(II)
    Assay 99%

    GHS H Statement
    H350-H317
    May cause cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit