Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dibromochloromethane, 98%

Click to view available options
Quantity:
5g
25g
100g
124-48-1
CHBr2Cl
208.277
MFCD00000820
GATVIKZLVQHOMN-UHFFFAOYSA-N
chlorodibromomethane,methane, dibromochloro,dibromo chloro methane,monochlorodibromomethane,cdbm,methane, chlorodibromo,unii-3t4ajr1h24,ccris 938,dibromo-chloro-methane,chlorobromoform
31296
CHEBI:34627
dibromo(chloro)methane
C(Cl)(Br)Br
124-48-1
CHBr2Cl
208.277
MFCD00000820
GATVIKZLVQHOMN-UHFFFAOYSA-N
chlorodibromomethane,methane, dibromochloro,dibromo chloro methane,monochlorodibromomethane,cdbm,methane, chlorodibromo,unii-3t4ajr1h24,ccris 938,dibromo-chloro-methane,chlorobromoform
31296
CHEBI:34627
dibromo(chloro)methane
C(Cl)(Br)Br
Applications
It affords bromochlorocarbene under phase-transfer conditions.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture & Light Sensitive. Store away from light, water/moisture, oxidizing agents and heat. Store under inert gas.
Density 2.42
Melting Point -22°C
Refractive Index 1.5475
Sensitivity Light and moisture sensitive
Boiling Point 115°C to 118°C
Quantity 100g
Beilstein 1731046
Merck Index 14,2135
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 208.28
Percent Purity 98%
Chemical Name or Material Dibromochloromethane
Show More Show Less

GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 204-704-0

RTECSNumber : PA6360000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen