Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dibromomalonamide, 98+%

Click to view available options
Quantity:
10g
25g
100g
73003-80-2
C3H4Br2N2O2
259.885
MFCD00014803
SWHQVMGRXIYDSF-UHFFFAOYSA-N
dibromomalonamide,2,2-dibromomalonamide,dibromopropanediamide,propanediamide, 2,2-dibromo,dibromo malonamide-13c3,dibromo malonamide,dibromomalonic diamide,dibromomalonic acid diamide,propanediamide,2,2-dibromo,2,2-bis bromanyl propanediamide
175375
2,2-dibromopropanediamide
C(=O)(C(C(=O)N)(Br)Br)N
73003-80-2
C3H4Br2N2O2
259.885
MFCD00014803
SWHQVMGRXIYDSF-UHFFFAOYSA-N
dibromomalonamide,2,2-dibromomalonamide,dibromopropanediamide,propanediamide, 2,2-dibromo,dibromo malonamide-13c3,dibromo malonamide,dibromomalonic diamide,dibromomalonic acid diamide,propanediamide,2,2-dibromo,2,2-bis bromanyl propanediamide
175375
2,2-dibromopropanediamide
C(=O)(C(C(=O)N)(Br)Br)N
Applications
Dibromomalonamide is an antimicrobial agent used in sugar mills, a compound for lipophilic fluid systems for fabric care. It is also used to produce 5-thioxo-[1,3]dithiolo[4,5-d][1,3]dithiole-2,2-dicarboxylic acid diamide.

Solubility
Soluble in water.

Notes
Moisture & light sensitive. Store away from oxidizing agent.
Melting Point 202°C to 207°C
Quantity 25g
Beilstein 1770873
Formula Weight 259.89
Percent Purity ≥98%
Chemical Name or Material Dibromomalonamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA