Alfa Aesar™ Dibromomethane, 99%, stab. with 50ppm BHT

Manufacturer: Alfa Aesar™ A1045636

Quantity 500g

 View more versions of this product

Catalog No. AAA1045636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 74-95-3
    Molecular Formula CH2Br2
    Formula Weight 173.85
    MDL Number MFCD00000168
    UN Number UN2664
    Beilstein 969143
    Merck Index 14,6061
    Synonym Methylene bromide
    Chemical Name or Material Dibromomethane, Stabilized with 50ppm BHT

    Specifications

    Formula Weight 173.85
    Health Hazard 3 GHS P Statement
    P261-P271-P304+P340-P312
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use only outdoors or in a well-ventilated area.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    CAS 74-95-3
    UN Number UN2664
    Beilstein 969143
    Synonym Methylene bromide
    Health Hazard 2 GHS H Statement
    H332
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DIBROMOMETHANE
    Melting Point −53°C
    Boiling Point 96° to 98°C
    Molecular Formula CH2Br2
    Merck Index 14,6061
    Health Hazard 1 Warning
    Quantity 500g
    MDL Number MFCD00000168
    RTECS Number PA7350000
    TSCA TSCA
    Chemical Name or Material Dibromomethane, Stabilized with 50ppm BHT
    Assay 99%

    Description

    Applications
    Dibromomethane is used as solvent in organic synthesis. It acts as an intermediate in the manufacture of specialty chemicals, agrochemicals and pharmaceuticals. It is useful as extractant and utilized for the determination of 5-nitroimidazoles (5-NDZ) in environmental waters. It is involved in the convertion of catechols to their methylenedioxy derivatives.

    Solubility
    Miscible with water, chloroform, acetone, ether and alcohol.

    Notes
    Store in cool place. Incompatible with strong oxidizing agents, aluminum and magnesium.

    GHS H Statement
    H332
    Harmful if inhaled.

    GHS P Statement
    P261-P271-P304+P340-P312
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use only outdoors or in a well-ventilated area.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit