Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibucaine, USP, 97-102.5%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
85-79-0
C20H29N3O2
343.47
PUFQVTATUTYEAL-UHFFFAOYSA-N
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide
CCCCOC1=CC(C(=O)NCCN(CC)CC)=C2C=CC=CC2=N1
85-79-0
C20H29N3O2
343.47
PUFQVTATUTYEAL-UHFFFAOYSA-N
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide
CCCCOC1=CC(C(=O)NCCN(CC)CC)=C2C=CC=CC2=N1

DI113, 85-79-0, C20H29N3O2

Spectrum™ Dibucaine, USP is used to treat minor skin irritations like pain and itching is also used to relieve rectal pain and itching caused by hemorrhoids or other rectal irritations. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.002
Quantity 5 g
Loss on Drying 0.01
Melting Range 62.5-66.0°C
CAS Max % 1
UN Number UN1544
Percent Purity 97 to 102.5%