Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl maleate, 97%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
105-76-0
C12H20O4
228.29
MFCD00009447
JBSLOWBPDRZSMB-FPLPWBNLSA-N
dibutyl maleate,butyl maleate,staflex dbm,rc comonomer dbm,maleic acid, dibutyl ester,di-n-butyl maleate,2-butenedioic acid z-, dibutyl ester,2-butenedioic acid, dibutyl ester,maleic acid dibutyl ester,2-butenedioic acid 2z-, dibutyl ester
5271569
dibutyl (Z)-but-2-enedioate
CCCCOC(=O)C=CC(=O)OCCCC
105-76-0
C12H20O4
228.29
MFCD00009447
JBSLOWBPDRZSMB-FPLPWBNLSA-N
dibutyl maleate,butyl maleate,staflex dbm,rc comonomer dbm,maleic acid, dibutyl ester,di-n-butyl maleate,2-butenedioic acid z-, dibutyl ester,2-butenedioic acid, dibutyl ester,maleic acid dibutyl ester,2-butenedioic acid 2z-, dibutyl ester
5271569
dibutyl (Z)-but-2-enedioate
CCCCOC(=O)C=CC(=O)OCCCC
Boiling Point 274°C to 277°C
Flash Point 120°C
Specific Gravity 0.99
Density 0.9900g/mL
Refractive Index 1.4440 to 1.446
Quantity 1kg
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)3OCOCHCHCOO(CH2)3CH3
Beilstein 02, IV, 2209
Solubility Information Solubility in water: insoluble
Formula Weight 228.29
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Dibutyl maleate, 97%
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-328-4

RTECSNumber : ON0875000

TSCA : TSCA