Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl succinate, 99%, Acros Organics

Click to view available options
Quantity:
5mL
25mL
100mL
141-03-7
C12H22O4
230.3
YUXIBTJKHLUKBD-UHFFFAOYSA-N
dibutyl succinate,tabutrex,succinic acid di-n-butyl ester,di-n-butyl succinate,tabatrex,dibutylsuccinate,butyl butanedioate,butanedioic acid, dibutyl ester,succinic acid dibutyl ester,dnbs
8830
CHEBI:82263
dibutyl butanedioate
CCCCOC(=O)CCC(=O)OCCCC
141-03-7
C12H22O4
230.3
YUXIBTJKHLUKBD-UHFFFAOYSA-N
dibutyl succinate,tabutrex,succinic acid di-n-butyl ester,di-n-butyl succinate,tabatrex,dibutylsuccinate,butyl butanedioate,butanedioic acid, dibutyl ester,succinic acid dibutyl ester,dnbs
8830
CHEBI:82263
dibutyl butanedioate
CCCCOC(=O)CCC(=O)OCCCC
Boiling Point 274.0°C
Flash Point 144°C
Specific Gravity 0.98
Quantity 25mL
Density 0.9800g/mL
Melting Point -29.0°C
Assay Percent Range 98.5% min. (GC)
Linear Formula C4H9O2CH2CH2CO2C4H9
Solubility Information Solubility in water: slightly.
Formula Weight 230.3
Percent Purity 99%
Chemical Name or Material Dibutyl succinate
Show More Show Less

EINECSNumber : 205-449-8