Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutylamine, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
111-92-2
C8H19N
129.247
JQVDAXLFBXTEQA-UHFFFAOYSA-N
8148
N-butylbutan-1-amine
CCCCNCCCC
111-92-2
C8H19N
129.247
JQVDAXLFBXTEQA-UHFFFAOYSA-N
8148
N-butylbutan-1-amine
CCCCNCCCC

High quality chemicals for laboratory and specialized industrial use

Packaging Narrow Mouth Amber Glass Bottle
Quantity 500mL
Flash Point 47.00°C
Specific Gravity 1L = 0.76kg
Formula Weight 129.25
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Dibutylamine