Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dibutylamine, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Specifications
Specifications
Flash Point | 47.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 500 mL |
Specific Gravity | 1L = 0.76kg |
Formula Weight | 129.25 |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Dibutylamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction