Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutylamine, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
111-92-2
C8H19N
129.247
JQVDAXLFBXTEQA-UHFFFAOYSA-N
dibutylamine,di-n-butylamine,1-butanamine, n-butyl,n-butyl-1-butanamine,dibutilamina,n-dibutylamine,di-n-butyl amine,dibutyl amine,di-normal-butylamine,dibutyl-amine
8148
N-butylbutan-1-amine
CCCCNCCCC
111-92-2
C8H19N
129.247
JQVDAXLFBXTEQA-UHFFFAOYSA-N
dibutylamine,di-n-butylamine,1-butanamine, n-butyl,n-butyl-1-butanamine,dibutilamina,n-dibutylamine,di-n-butyl amine,dibutyl amine,di-normal-butylamine,dibutyl-amine
8148
N-butylbutan-1-amine
CCCCNCCCC

High quality chemicals for laboratory and specialized industrial use

Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.76kg
Quantity 4L
Flash Point 47.00°C
Formula Weight 129.25
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Dibutylamine