Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyltin dichloride, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
1kg
Packaging:
Glass bottle
683-18-1
C8H18Cl2Sn
303.842
MFCD00000518
RJGHQTVXGKYATR-UHFFFAOYSA-L
dibutyltin dichloride,dibutyldichlorotin,di-n-butyltin dichloride,stannane, dibutyldichloro,dibutyltin chloride,dichlorodibutyltin,dibutyl dichloro stannane,dichlorodibutylstannane,dibutyltindichloride,dibutyl tin dichloride
12688
dibutyl(dichloro)stannane
CCCC[Sn](CCCC)(Cl)Cl
683-18-1
C8H18Cl2Sn
303.842
MFCD00000518
RJGHQTVXGKYATR-UHFFFAOYSA-L
dibutyltin dichloride,dibutyldichlorotin,di-n-butyltin dichloride,stannane, dibutyldichloro,dibutyltin chloride,dichlorodibutyltin,dibutyl dichloro stannane,dichlorodibutylstannane,dibutyltindichloride,dibutyl tin dichloride
12688
dibutyl(dichloro)stannane
CCCC[Sn](CCCC)(Cl)Cl
Density 1.4000g/mL
Specific Gravity 1.4
Packaging Glass bottle
Flash Point 112°C
Infrared Spectrum Authentic
Quantity 250g
Color White to Beige
Boiling Point 135.0°C (10.0 mmHg)
Melting Point 39.0°C to 41.0°C
Assay Percent Range 97%
Linear Formula [CH3(CH2)3]2SnCl2
Fieser 01,213; 01,1294; 05,622
Formula Weight 303.83
Physical Form Crystalline Low Melting Mass
Percent Purity 97%
Chemical Name or Material Dibutyltin dichloride
Show More Show Less

GHS H Statement
Very toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. Suspected of causing genetic defects. Causes damage to organs through prolonged or repeated exposure. Harmful in contact with skin. Fatal if inhaled. Toxic if swallowed. May damage fertility. May damage the unborn child.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.

Danger