Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dibutyltin dilaurate, 94%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 205.0°C (12.0 mmHg) |
Melting Point | 16.0°C to 18.0°C |
Color | Yellow |
Physical Form | Oily Liquid |
Quantity | 5 g |
Assay Percent Range | 94% |
Linear Formula | [CH3(CH2)3]2Sn[OC(O)CH2(CH2)9CH3]2 |
Flash Point | 149°C |
Merck Index | 15,3047 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in ether and aceton |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause an allergic skin reaction. Causes damage to organs if swallowed. May damage fertility. May damage the unborn child. Suspected of causing genetic defect |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. |
EINECSNumber | 201-039-8 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'