Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyltin oxide, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
818-08-6
C8H18OSn
248.92
MFCD00001992
JGFBRKRYDCGYKD-UHFFFAOYSA-N
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
61221
dibutyl(oxo)tin
CCCC[Sn](=O)CCCC
818-08-6
C8H18OSn
248.92
MFCD00001992
JGFBRKRYDCGYKD-UHFFFAOYSA-N
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
61221
dibutyl(oxo)tin
CCCC[Sn](=O)CCCC
Chloride 0.2% max
Melting Point >300.0°C
Quantity 500g
Color White
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula [CH3(CH2)3]2Sn(=O)
Beilstein 04, I, 588
Fieser 05,188; 09,141; 10,123; 12,160; 13,53; 15,116; 16,112
Merck Index 15, 3048
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in organic solvents
Formula Weight 248.92
Physical Form Powder or Granular Powder
Percent Purity 98%
Chemical Name or Material Dibutyltin oxide, 98%
Show More Show Less

GHS H Statement
May damage fertility or the unborn child.
Toxic if swallowed.
Suspected of causing genetic defects.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure.
Very toxic

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa

GHS Signal Word: Danger

EINECSNumber : 212-449-1

RTECSNumber : WH7175000

TSCA : TSCA