Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicarbonyl(pentamethylcyclopentadienyl)chromium dimer, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
37299-12-0
C24H34Cr2O4
490.52
MFCD00058843
AVZOFKYVNIQWMS-UHFFFAOYSA-N
pentamethylcyclopentadienylchromium dicarbonyl dimer,dicarbonyl pentamethylcyclopentadienyl chromium v dimer
37299-12-0
C24H34Cr2O4
490.52
MFCD00058843
AVZOFKYVNIQWMS-UHFFFAOYSA-N
pentamethylcyclopentadienylchromium dicarbonyl dimer,dicarbonyl pentamethylcyclopentadienyl chromium v dimer
Quantity 1g
Color Gray to Black
Packaging Glass bottle
Infrared Spectrum Authentic
Formula Weight 486.49
Physical Form Crystals
Chemical Name or Material Dicarbonyl(pentamethylcyclopentadienyl)chromium dimer

GHS P Statement
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger