Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dicarbonylacetylacetonato rhodium(I), Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 154.0°C to 156.0°C |
Color | Green-Red |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 500 mg |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 258.04 |
Physical Form | Flakes or Needles |
Chemical Name or Material | Dicarbonylacetylacetonato rhodium(I) |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Flammable solid. Toxic if swallowed. May cause an allergic skin reaction. Causes serious eye irritation. Harmful to aquatic life with long lasting effects. Risk of explosion if heated under confinement |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occ |
EINECSNumber | 238-947-9 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction