Alfa Aesar™ Dicarbonylbis(cyclopentadienyl)titanium(II), 98%

Manufacturer: Alfa Aesar™ 3957104

Quantity 2g

 View more versions of this product

Catalog No. AA3957104

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12129-51-0
    Formula Weight 234.09
    MDL Number MFCD00143762
    UN Number UN3466
    Chemical Name or Material Dicarbonylbis(cyclopentadienyl)titanium(II)

    Specifications

    Formula Weight 234.09
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P301+P310-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 12129-51-0
    MDL Number MFCD00143762
    Health Hazard 1 Danger
    Quantity 2g
    Physical Form Crystalline
    UN Number UN3466
    Boiling Point 60°C (0.001mmHg) (subl.)
    Melting Point 90°C (dec.)
    Health Hazard 2 GHS H Statement
    H228-H300-H331
    Flammable solid.
    Fatal if swallowed.
    Toxic if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    Sensitivity Air & Moisture Sensitive
    Chemical Name or Material Dicarbonylbis(cyclopentadienyl)titanium(II)
    Assay 98%

    GHS H Statement
    H228-H300-H331
    Flammable solid.
    Fatal if swallowed.
    Toxic if inhaled.

    GHS P Statement
    P210-P261-P280-P301+P310-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit