Alfa Aesar™ Dicarbonylcyclopentadienylcobalt

Catalyst for the cyclotrimerization of alkynes

Manufacturer: Alfa Aesar™ 2313606

Quantity 5g

 View more versions of this product

Catalog No. AA2313606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12078-25-0
    Formula Weight 180.05
    MDL Number MFCD00009589
    UN Number UN3281
    Chemical Name or Material Dicarbonylcyclopentadienylcobalt

    Specifications

    Formula Weight 180.05
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00009589
    UN Number UN3281
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H226-H341-H317
    Fatal if swallowed.
    Flammable liquid and vapor.
    Suspected of causing genetic defects.
    May cause an allergic skin reaction.
    Sensitivity Air Sensitive
    Physical Form Liquid
    CAS 12078-25-0
    Boiling Point 139° to 140°C (710mmHg)
    Flash Point 26° (80°F)
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, LIQUID, N.O.S.
    Quantity 5g
    Chemical Name or Material Dicarbonylcyclopentadienylcobalt

    GHS H Statement
    H300-H226-H341-H317
    Fatal if swallowed.
    Flammable liquid and vapor.
    Suspected of causing genetic defects.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit