Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicarbonylcyclopentadienylruthenium(II) dimer, Acros Organics

Click to view available options
Quantity:
250mg
1g
12132-87-5
C14H10O4Ru2
12132-87-5
C14H10O4Ru2
Quantity 1g
Packaging Glass bottle
Formula Weight 444.37
Chemical Name or Material Dicarbonylcyclopentadienylruthenium(II) dimer