Learn More
Dicer Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™
SDP
Mouse Recombinant Monoclonal Antibody
Supplier:  Active Motif 91242
Encompass
 View more versions of this product
Catalog No. 50-199-3766
Add to Cart
Description
Description
Sphingosine Kinase (SphK) catalyzes the phosphorylation of the lipid sphingosine, creating the bioactive lipid sphingosine-1-phosphate (S1P). S1P subsequently signals through cell surface G protein-coupled receptors, as well as intracellularly, to modulate cell proliferation, survival, motility and differentiation. SphK is an important signaling enzyme which is activated by diverse agents, including growth factors that signal through receptor tyrosine kinases, agents activating G protein-coupled receptors, and immunoglobulin receptors. Two SphK isotypes, SphK-1 and SphK-2, have been cloned, and both isotypes are ubiquitously expressed. SphK-1 has been shown to mediate cell growth, prevention of apoptosis, and cellular transformation, and is upregulated in a variety of human tumors. In contrast, SphK-2 increases apoptosis, and may be responsible for phosphorylating and activating the immunosuppressive drug FTY720.
Specifications
Specifications
Dicer
Monoclonal
Unconjugated
DICER1
1110006F08Rik; D12Ertd7e; DCR; DCR1; DCR-1; DICER; dicer 1 ribonuclease III; dicer 1, double-stranded RNA-specific endoribonuclease; dicer 1, ribonuclease III; dicer 1, ribonuclease type III; DICER1; Dicer1, Dcr-1 homolog; Dicer1e; double-strand-specific ribonuclease mDCR-1; Endoribonuclease Dicer; Helicase MOI; Helicase with RNase motif; helicase-moi; HERNA; K12H4.8-LIKE; KIAA0928; mKIAA0928; MNG1; RMSE2
Mouse
Protein A
RUO
192119
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5
Q8R418
DICER1
This antibody was raised against amino acids 1638-1899 of mouse Dicer (aka dicer 1, ribonuclease type III).
10 μg
Primary
Mouse
Antibody
IgG2a