Learn More
Dicer Mouse anti-Human, Mouse, Rat, Alexa Fluor 700, Clone: S167-7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP222415AF700
Catalog No. NB222415700
Description
Description
Dicer Monoclonal specifically detects Dicer in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Dicer
S167-7
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
DICER1
Fusion protein amino acids 1638-1899 of mouse Endoribonuclease Dicer
Protein G purified
Apoptosis, Cell Biology, Cell Cycle and Replication, Chromatin Research, Cytokine Research, Epigenetics, Immunology, Innate Immunity, microRNA, Signal Transduction
Primary
23405
Human, Mouse, Rat
Western Blot, Immunocytochemistry, Immunofluorescence
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
DCR1, Dicer, dicer 1, double-stranded RNA-specific endoribonuclease, dicer 1, ribonuclease type III, Dicer1, Dcr-1 homolog (Drosophila), EC 3.1.26, EC 3.1.26.-, EC 3.1.26.3, Helicase MOI, Helicase with RNase motif, helicase-moi, HERNADicer1, Dcr-1 homolog, K12H4.8-LIKE, KIAA0928endoribonuclease Dicer
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Detects approx 215kDa.